missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxovalerate 97.0+%, TCI America™

Catalog No. p-7131750
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00315G 5 g
K003125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00315G Supplier TCI America Supplier No. K00315G

Chemical Identifiers

CAS 4949-44-4
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009317
InChI Key UDRCONFHWYGWFI-UHFFFAOYSA-N
Synonym ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate
PubChem CID 78656
IUPAC Name ethyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OCC

Specifications

Color Colorless
Boiling Point 191°C
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Oxovalerate
EINECSNumber (2)-1505
TSCA No