missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxohexanoate 95.0+%, TCI America™

Supplier:  TCI America K0030500ML

 View more versions of this product

Catalog No. K0030500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-Oxohexanoate
-44°C
95°C
MFCD00009401
ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
CCCC(=O)CC(=O)OCC
158.20
CHEBI:18119
≥95.0% (GC)
3249-68-1
Yellow
C8H14O3
500 mL
KQWWVLVLVYYYDT-UHFFFAOYSA-N
ethyl 3-oxohexanoate
238498
158.20
Liquid

Chemical Identifiers

3249-68-1
158.20
KQWWVLVLVYYYDT-UHFFFAOYSA-N
238498
ethyl 3-oxohexanoate
C8H14O3
MFCD00009401
ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
CHEBI:18119
CCCC(=O)CC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1505

RTECSNumber : MO7420500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions