missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Oxoheptanoate 95.0+%, TCI America™
$322.48 - $542.44
Chemical Identifiers
| CAS | 7737-62-4 |
|---|---|
| Molecular Formula | C9H16O3 |
| Molecular Weight (g/mol) | 172.22 |
| MDL Number | MFCD00143188 |
| InChI Key | UKRVECBFDMVBPU-UHFFFAOYSA-N |
| Synonym | Ethyl 3-Oxoenanthate, 3-Oxoheptanoic Acid Ethyl Ester, 3-Ketoheptanoic Acid Ethyl Ester, 3-Oxoenanthic Acid Ethyl Ester |
| PubChem CID | 559036 |
| IUPAC Name | ethyl 3-oxoheptanoate |
| SMILES | CCCCC(=O)CC(=O)OCC |
Chemical Identifiers
| 7737-62-4 | |
| 172.22 | |
| UKRVECBFDMVBPU-UHFFFAOYSA-N | |
| 559036 | |
| CCCCC(=O)CC(=O)OCC |
| C9H16O3 | |
| MFCD00143188 | |
| Ethyl 3-Oxoenanthate, 3-Oxoheptanoic Acid Ethyl Ester, 3-Ketoheptanoic Acid Ethyl Ester, 3-Oxoenanthic Acid Ethyl Ester | |
| ethyl 3-oxoheptanoate |
Specifications
| 7737-62-4 | |
| 111°C | |
| MFCD00143188 | |
| Ethyl 3-Oxoenanthate, 3-Oxoheptanoic Acid Ethyl Ester, 3-Ketoheptanoic Acid Ethyl Ester, 3-Oxoenanthic Acid Ethyl Ester | |
| CCCCC(=O)CC(=O)OCC | |
| 172.22 | |
| 172.22 | |
| Liquid |
| Colorless | |
| C9H16O3 | |
| 10 g | |
| UKRVECBFDMVBPU-UHFFFAOYSA-N | |
| ethyl 3-oxoheptanoate | |
| 559036 | |
| ≥95.0% (GC) | |
| Ethyl 3-Oxoheptanoate |
Safety and Handling
TSCA : No