missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxoheptanoate 95.0+%, TCI America™

Catalog No. O021610G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O021610G 10 g
O021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O021610G Supplier TCI America Supplier No. O021610G

Chemical Identifiers

CAS 7737-62-4
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00143188
InChI Key UKRVECBFDMVBPU-UHFFFAOYSA-N
Synonym Ethyl 3-Oxoenanthate, 3-Oxoheptanoic Acid Ethyl Ester, 3-Ketoheptanoic Acid Ethyl Ester, 3-Oxoenanthic Acid Ethyl Ester
PubChem CID 559036
IUPAC Name ethyl 3-oxoheptanoate
SMILES CCCCC(=O)CC(=O)OCC

Specifications

Color Colorless
Boiling Point 111°C
Quantity 10 g
Formula Weight 172.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Oxoheptanoate