missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxo-3-(4-pyridyl)propionate 98.0+%, TCI America™

Catalog No. p-7129292
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10321G 1 g
E10325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10321G Supplier TCI America Supplier No. E10321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26377-17-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094021
InChI Key PCJNYGPKMQQCPX-UHFFFAOYSA-N
Synonym ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
PubChem CID 2735202
IUPAC Name ethyl 3-oxo-3-(pyridin-4-yl)propanoate
SMILES CCOC(=O)CC(=O)C1=CC=NC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 169°C
Quantity 1 g
Formula Weight 193.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Oxo-3-(4-pyridyl)propionate