missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methylisoxazole-4-carboxylate, 97%

Catalog No. p-7046799
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1636203 1 g
AAL1636206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1636203 Supplier Thermo Scientific Chemicals Supplier No. L1636203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20328-15-8 | C7H9NO3 | 155.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20328-15-8
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD01075736
InChI Key AEWHICNMVQOWJE-UHFFFAOYSA-N
Synonym ethyl 3-methylisoxazole-4-carboxylate, ethyl 3-methyl-isoxazole-4-carboxylate, 4-isoxazolecarboxylic acid, 3-methyl-, ethyl ester, acmc-20akze, ethyl 3-methyl-4-isoxazolecarboxylate, 3-methyl-4-isoxazolecarboxylic acid ethyl ester, 3-methyl-isoxazole-4-carboxylic acid ethyl ester, 4-isoxazolecarboxylicacid, 3-methyl-, ethyl ester, 3-methyl-4-isoxazolecarboxylic acid, ethyl ester, 3-methylisoxazole-4-carboxylic acid ethyl ester
PubChem CID 317668
IUPAC Name ethyl 3-methyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=CON=C1C

Specifications

Boiling Point 64°C (0.6 mmHg)
Refractive Index 1.4595
Quantity 1 g
Beilstein 1072756
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material Ethyl 3-methylisoxazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only