missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers) 95.0+%, TCI America™

Catalog No. E087325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E087325G 25 g
E0873250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E087325G Supplier TCI America Supplier No. E087325G

Chemical Identifiers

CAS 77-83-8
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00022338
InChI Key LQKRYVGRPXFFAV-UHFFFAOYSA-N
Synonym ethyl methylphenylglycidate, ethyl 3-methyl-3-phenylglycidate, strawberry aldehyde, fraeseol, empg, c-16 aldehyde, aldehyde c-16, ethyl methylphenyl glycidate, ethyl-3-methyl-3-phenylglycidate, ethyl 2,3-epoxy-3-phenylbutyrate
PubChem CID 6501
IUPAC Name ethyl 3-methyl-3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)(C)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers)
EINECSNumber (3)-3096
RTECSNumber MW5250000
TSCA Yes