missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Methoxypropionate 99.0+%, TCI America™

Catalog No. M100110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M100110G 10 g
M100125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M100110G Supplier TCI America Supplier No. M100110G
Only null left

Chemical Identifiers

CAS 10606-42-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00143172
InChI Key IJUHLFUALMUWOM-UHFFFAOYSA-N
Synonym 3-Methoxypropionic Acid Ethyl Ester
PubChem CID 82767
IUPAC Name ethyl 3-methoxypropanoate
SMILES CCOC(=O)CCOC

Specifications

Color Colorless
Boiling Point 158°C
Quantity 10 g
UN Number 3272
Formula Weight 132.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Methoxypropionate
EINECSNumber (2)-1350&(2)-1379
RTECSNumber UF5273000
TSCA No