missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Mercaptopropionate 98.0+%, TCI America™

Catalog No. M1038500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1038500G 500 g
M103825G 25 g
M1038100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1038500G Supplier TCI America Supplier No. M1038500G

Chemical Identifiers

CAS 5466-06-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00004896
InChI Key CJQWLNNCQIHKHP-UHFFFAOYSA-N
Synonym ethyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, ethyl ester, 3-mercaptopropanoic acid ethyl ester, 3-mercaptopropionic acid ethyl ester, ethyl 3-mercaptopropanoate, ethyl beta-mercaptopropionate, unii-22gqz8p70d, propionic acid, 3-mercapto-, ethyl ester, ethyl 3-mercaptopropanoic acid, ethyl .beta.-mercaptopropionate
PubChem CID 21625
IUPAC Name ethyl 3-sulfanylpropanoate
SMILES CCOC(=O)CCS

Specifications

Color Colorless
Quantity 500 g
Formula Weight 134.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Mercaptopropionate