missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific™

Catalog No. AC319270010
Change view
Click to view available options
Glass bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC319270010 Glass bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC319270010 Supplier Thermo Scientific Chemicals Supplier No. 319270010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5100-34-5
Density 1.0870g/mL
Boiling Point 68.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H9NO3
Refractive Index 1.4275 to 1.4295
Linear Formula OCNCH2CH2C(=O)OCH2CH3
Quantity 1g
MDL Number MFCD00078461
Specific Gravity 1.087
Synonym ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
InChI Key XBSGYVHOINMTIM-UHFFFAOYSA-N
SMILES CCOC(=O)CCN=C=O
IUPAC Name ethyl 3-isocyanatopropanoate
Molecular Weight (g/mol) 143.14
PubChem CID 2733373
Formula Weight 143.07
Percent Purity 98%
Chemical Name or Material Ethyl 3-isocyanatopropionate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only