missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isocyanatopropionate, 98%

Catalog No. AAL0961603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0961603 1 g
AAL0961606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0961603 Supplier Thermo Scientific Chemicals Supplier No. L0961603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5100-34-5 | C6H9NO3 | 143.14 g/mol

Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

Solubility
Not miscible with water.

Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.

Chemical Identifiers

CAS 5100-34-5
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD00078461
InChI Key XBSGYVHOINMTIM-UHFFFAOYSA-N
Synonym ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
PubChem CID 2733373
IUPAC Name ethyl 3-isocyanatopropanoate
SMILES CCOC(=O)CCN=C=O

Specifications

Density 1.087
Boiling Point 74°C to 75°C (4 mmHg)
Flash Point 92°C (197°F)
Odor Irritating
Refractive Index 1.429
Quantity 1 g
UN Number UN2206
Beilstein 1705618
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 143.14
Percent Purity 98%
Chemical Name or Material Ethyl 3-isocyanatopropionate
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only