missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxyhexanoate, 97%

CAS: 2305-25-1 | C8H16O3 | 160.213 g/mol

$105.75 - $346.00

Chemical Identifiers

CAS 2305-25-1
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00036604
InChI Key LYRIITRHDCNUHV-UHFFFAOYSA-N
Synonym hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
PubChem CID 61293
ChEBI CHEBI:23997
IUPAC Name ethyl 3-hydroxyhexanoate
SMILES CCCC(CC(=O)OCC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1206014
View Documents
Thermo Scientific Chemicals
A1206014
25 g
Each for $105.75
Only null left
 
AAA1206022
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1206022
100 g
Each for $346.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2305-25-1
160.213
LYRIITRHDCNUHV-UHFFFAOYSA-N
61293
ethyl 3-hydroxyhexanoate
C8H16O3
MFCD00036604
hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
CHEBI:23997
CCCC(CC(=O)OCC)O

Specifications

2305-25-1
90°C to 92°C (14 mmHg)
C8H16O3
1.4298
25 g
LYRIITRHDCNUHV-UHFFFAOYSA-N
ethyl 3-hydroxyhexanoate
61293
160.21
Ethyl 3-hydroxyhexanoate
0.974
94°C (202°F)
CH3CH2CH2CHOHCH2CO2CH2CH3
MFCD00036604
hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
CCCC(CC(=O)OCC)O
160.213
CHEBI:23997
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-973-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only