missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Furancarboxylate 98.0+%, TCI America™
$104.38 - $301.98
Chemical Identifiers
| CAS | 614-98-2 |
|---|---|
| Molecular Formula | C7H8O3 |
| Molecular Weight (g/mol) | 140.138 |
| MDL Number | MFCD00005348 |
| InChI Key | LOFDXZJSDVCYAS-UHFFFAOYSA-N |
| PubChem CID | 69201 |
| IUPAC Name | ethyl furan-3-carboxylate |
| SMILES | CCOC(=O)C1=COC=C1 |
Chemical Identifiers
| 614-98-2 | |
| 140.138 | |
| LOFDXZJSDVCYAS-UHFFFAOYSA-N | |
| ethyl furan-3-carboxylate |
| C7H8O3 | |
| MFCD00005348 | |
| 69201 | |
| CCOC(=O)C1=COC=C1 |
Specifications
| 614-98-2 | |
| 95°C | |
| MFCD00005348 | |
| 3272 | |
| LOFDXZJSDVCYAS-UHFFFAOYSA-N | |
| ethyl furan-3-carboxylate | |
| 69201 | |
| ≥98.0% (GC) | |
| Ethyl 3-Furancarboxylate |
| Yellow | |
| C7H8O3 | |
| 5 g | |
| ethyl 3-furoate, ethyl 3-furancarboxylate, unii-i8q0yn919k, 3-furancarboxylic acid, ethyl ester, 3-furancarboxylic acid ethyl ester, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, acmc-1bczk | |
| CCOC(=O)C1=COC=C1 | |
| 140.138 | |
| 140.14 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator