missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Ethoxypropionate 99.0+%, TCI America™

Catalog No. p-7128932
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E031925ML 25 mL
E0319500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E031925ML Supplier TCI America Supplier No. E031925ML

Chemical Identifiers

CAS 763-69-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD00051356
InChI Key BHXIWUJLHYHGSJ-UHFFFAOYSA-N
Synonym ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
PubChem CID 12989
IUPAC Name ethyl 3-ethoxypropanoate
SMILES CCOCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 mL
UN Number 3272
Formula Weight 146.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Ethoxypropionate
RTECSNumber UF3325000
TSCA Yes