missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Ethoxyacrylate (cis- and trans- mixture) 98.0+%, TCI America™
$92.41 - $306.10
Chemical Identifiers
| CAS | 1001-26-9 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00009863 |
| InChI Key | ITQFPVUDTFABDH-AATRIKPKSA-N |
| PubChem CID | 5366269 |
| IUPAC Name | ethyl (E)-3-ethoxyprop-2-enoate |
| SMILES | CCOC=CC(=O)OCC |
Chemical Identifiers
| 1001-26-9 | |
| 144.17 | |
| ITQFPVUDTFABDH-AATRIKPKSA-N | |
| ethyl (E)-3-ethoxyprop-2-enoate |
| C7H12O3 | |
| MFCD00009863 | |
| 5366269 | |
| CCOC=CC(=O)OCC |
Specifications
| 1001-26-9 | |
| C7H12O3 | |
| 5 mL | |
| ITQFPVUDTFABDH-AATRIKPKSA-N | |
| ethyl (E)-3-ethoxyprop-2-enoate | |
| 5366269 | |
| ≥98.0% (GC) | |
| Ethyl 3-Ethoxyacrylate (cis- and trans- mixture) |
| Yellow | |
| MFCD00009863 | |
| ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate | |
| CCOC=CC(=O)OCC | |
| 144.17 | |
| 144.17 | |
| Liquid |
Safety and Handling
TSCA : No