missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Ethoxy-2,2-difluoro-3-hydroxypropionate 83.0+%, TCI America™

Catalog No. E05471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05471G Supplier TCI America Supplier No. E05471G
Only null left

Chemical Identifiers

CAS 141546-97-6
Molecular Formula C7H12F2O4
Molecular Weight (g/mol) 198.166
MDL Number MFCD00718642
InChI Key DCDGEVPPIDIMON-UHFFFAOYSA-N
Synonym 3-Ethoxy-2,2-difluoro-3-hydroxypropionic Acid Ethyl Ester
PubChem CID 2758839
IUPAC Name ethyl 3-ethoxy-2,2-difluoro-3-hydroxypropanoate
SMILES CCOC(C(C(=O)OCC)(F)F)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 198.17
Percent Purity ≥83.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Ethoxy-2,2-difluoro-3-hydroxypropionate
TSCA No
Recommended Storage Refrigerator