missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-dimethylaminopropionate, 97%

Catalog No. AAL0154214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0154214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0154214 Supplier Thermo Scientific Chemicals Supplier No. L0154214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20120-21-2 | C7H15NO2 | 145.20 g/mol

Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20120-21-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.20
MDL Number MFCD00015166
InChI Key IZMJWGJGVGRWBW-UHFFFAOYSA-N
Synonym ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate
PubChem CID 88372
IUPAC Name ethyl 3-(dimethylamino)propanoate
SMILES CCOC(=O)CCN(C)C

Specifications

Density 0.915
Boiling Point 56°C to 57°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Amine-like
Refractive Index 1.419
Quantity 25 g
UN Number UN1993
Beilstein 1702477
Solubility Information Immiscible with water.
Formula Weight 145.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-dimethylaminopropionate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 243-524-7
RTECSNumber AY5992700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only