missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(Dimethylamino)acrylate 98.0+%, TCI America™
Supplier: TCI America D2836100G
Specifications
| Ethyl 3-(Dimethylamino)acrylate | |
| 19°C | |
| 136°C | |
| MFCD00144269 | |
| ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e | |
| CCOC(=O)C=CN(C)C | |
| 143.186 | |
| 143.19 | |
| Liquid |
| 924-99-2 | |
| Yellow | |
| C7H13NO2 | |
| 100 g | |
| MVUMJYQUKKUOHO-AATRIKPKSA-N | |
| ethyl (E)-3-(dimethylamino)prop-2-enoate | |
| 5369162 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 924-99-2 | |
| 143.186 | |
| MVUMJYQUKKUOHO-AATRIKPKSA-N | |
| 5369162 | |
| CCOC(=O)C=CN(C)C |
| C7H13NO2 | |
| MFCD00144269 | |
| ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e | |
| ethyl (E)-3-(dimethylamino)prop-2-enoate |
Safety and Handling
EINECSNumber : (2)-3661
TSCA : No