missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromopyruvate, tech. 75%

Catalog No. AAL0058206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0058206 5 g
AAL0058214 25 g
AAL0058222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0058206 Supplier Thermo Scientific Chemicals Supplier No. L0058206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol

Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

Solubility
Difficult to mix.

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 70-23-5
Molecular Formula C5H7BrO3
Molecular Weight (g/mol) 195.01
MDL Number MFCD00000204
InChI Key VICYTAYPKBLQFB-UHFFFAOYSA-N
Synonym ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
PubChem CID 66144
IUPAC Name ethyl 3-bromo-2-oxopropanoate
SMILES CCOC(=O)C(=O)CBr

Specifications

Density 1.555
Boiling Point 98°C to 100°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.469
Quantity 5 g
UN Number UN3265
Beilstein 1760158
Sensitivity Moisture sensitive
Solubility Information Difficult to mix.
Formula Weight 195.02
Percent Purity 75%
Grade Technical
Chemical Name or Material Ethyl 3-bromopyruvate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 200-729-6
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only