missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Bromoindole-2-carboxylate 95.0+%, TCI America™

Catalog No. p-7129227
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09271G 1 g
E09275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09271G Supplier TCI America Supplier No. E09271G
Only null left

Chemical Identifiers

CAS 91348-45-7
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD02071793
InChI Key DRJWEOYWZOGNQU-UHFFFAOYSA-N
Synonym ethyl 3-bromoindole-2-carboxylate, 3-bromoindole-2-carboxylic acid ethyl ester, 3-bromo-1h-indole-2-carboxylic acid ethyl ester, 3-bromo-2-indolecarboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 3-bromo-, ethyl ester, 3-br-ica-oet, ethyl 3-bromoindol-2-carboxylate, pubchem2247, pubchem7204, acmc-209r8t
PubChem CID 4715017
IUPAC Name ethyl 3-bromo-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=C(Br)C2=CC=CC=C2N1

Specifications

Melting Point 151°C
Color Yellow
Quantity 1 g
Formula Weight 268.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Bromoindole-2-carboxylate