missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6695106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6695106 5 g
AAH6695103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6695106 Supplier Thermo Scientific Chemicals Supplier No. H6695106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91348-45-7
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD02071793
InChI Key DRJWEOYWZOGNQU-UHFFFAOYSA-N
Synonym ethyl 3-bromoindole-2-carboxylate, 3-bromoindole-2-carboxylic acid ethyl ester, 3-bromo-1h-indole-2-carboxylic acid ethyl ester, 3-bromo-2-indolecarboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 3-bromo-, ethyl ester, 3-br-ica-oet, ethyl 3-bromoindol-2-carboxylate, pubchem2247, pubchem7204, acmc-209r8t
PubChem CID 4715017
IUPAC Name ethyl 3-bromo-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=C(Br)C2=CC=CC=C2N1

Specifications

Quantity 5 g
Beilstein 186192
Formula Weight 268.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 3-bromoindole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only