missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminopyrazole-4-carboxylate 98.0+%, TCI America™

Catalog No. p-7129137
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07665G 5 g
E076625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07665G Supplier TCI America Supplier No. E07665G
Only null left

Chemical Identifiers

CAS 6994-25-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00005238
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
PubChem CID 81472
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)N

Specifications

Melting Point 102°C
Color Yellow
Quantity 5 g
Formula Weight 155.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Aminopyrazole-4-carboxylate