missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98.5%

Catalog No. ac118031000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118031000 100 g Glass bottle
AC118035000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118031000 Supplier Thermo Scientific Chemicals Supplier No. 118031000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7318-00-5 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7318-00-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD02730138
InChI Key YPMPTULBFPFSEQ-PLNGDYQASA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Melting Point 33.0°C to 35.0°C
Color Yellow
Density 1.0220g/mL
Boiling Point 210.0°C to 215.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5000 to 1.5020
Linear Formula CH3C(NH2)=CHCO2C2H5
Quantity 100 g
Beilstein 03, 654
Specific Gravity 1.022
Formula Weight 129.16
Percent Purity 98.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Ethyl 3-aminocrotonate, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 230-782-0

RUO – Research Use Only