Learn More
Ethyl 3-aminocrotonate, 98.5%
CAS: 7318-00-5 | C6H11NO2 | 129.16 g/mol
$98.98 - $98.98
Chemical Identifiers
| CAS | 7318-00-5 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.16 |
| MDL Number | MFCD02730138 |
| InChI Key | YPMPTULBFPFSEQ-PLNGDYQASA-N |
| Synonym | z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate |
| PubChem CID | 643756 |
| IUPAC Name | ethyl (Z)-3-aminobut-2-enoate |
| SMILES | CCOC(=O)C=C(C)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118031000
|
Thermo Scientific Chemicals
118031000 |
100 g | Glass bottle |
Each for $98.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7318-00-5 | |
| 129.16 | |
| YPMPTULBFPFSEQ-PLNGDYQASA-N | |
| 643756 | |
| CCOC(=O)C=C(C)N |
| C6H11NO2 | |
| MFCD02730138 | |
| z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate | |
| ethyl (Z)-3-aminobut-2-enoate |
Specifications
| 7318-00-5 | |
| Yellow | |
| 210.0°C to 215.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5000 to 1.5020 | |
| MFCD02730138 | |
| 03, 654 | |
| z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate | |
| CCOC(=O)C=C(C)N | |
| 129.16 | |
| 129.16 | |
| Liquid or Low Melting Solid |
| 33.0°C to 35.0°C | |
| 1.0220g/mL | |
| 97°C | |
| 98% min. (GC) | |
| C6H11NO2 | |
| CH3C(NH2)=CHCO2C2H5 | |
| 100 g | |
| 1.022 | |
| YPMPTULBFPFSEQ-PLNGDYQASA-N | |
| ethyl (Z)-3-aminobut-2-enoate | |
| 643756 | |
| 98.5% | |
| Ethyl 3-aminocrotonate, 98.5% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 230-782-0
RUO – Research Use Only