missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Aminobenzofuran-2-carboxylate 98.0+%, TCI America™
$59.69 - $192.52
Chemical Identifiers
| CAS | 39786-35-1 |
|---|---|
| Molecular Formula | C11H11NO3 |
| Molecular Weight (g/mol) | 205.213 |
| MDL Number | MFCD00641330 |
| InChI Key | PWOARNMOPCOJEV-UHFFFAOYSA-N |
| Synonym | ethyl 3-aminobenzofuran-2-carboxylate, 3-aminocoumarilic acid ethyl ester, 2-benzofurancarboxylic acid, 3-amino-, ethyl ester, ethyl 3-aminobenzo b furan-2-carboxylate, ethyl 3-azanyl-1-benzofuran-2-carboxylate, 3-amino-2-benzofurancarboxylic acid ethyl ester, 3-aminobenzofuran-2-carboxylic acid ethyl ester, enamine_001390, ethyl 3-aminocoumarilate, ethyl3-aminobenzofuran-2-carboxylate |
| PubChem CID | 2063537 |
| IUPAC Name | ethyl 3-amino-1-benzofuran-2-carboxylate |
| SMILES | CCOC(=O)C1=C(C2=CC=CC=C2O1)N |
Chemical Identifiers
| 39786-35-1 | |
| 205.213 | |
| PWOARNMOPCOJEV-UHFFFAOYSA-N | |
| 2063537 | |
| CCOC(=O)C1=C(C2=CC=CC=C2O1)N |
| C11H11NO3 | |
| MFCD00641330 | |
| ethyl 3-aminobenzofuran-2-carboxylate, 3-aminocoumarilic acid ethyl ester, 2-benzofurancarboxylic acid, 3-amino-, ethyl ester, ethyl 3-aminobenzo b furan-2-carboxylate, ethyl 3-azanyl-1-benzofuran-2-carboxylate, 3-amino-2-benzofurancarboxylic acid ethyl ester, 3-aminobenzofuran-2-carboxylic acid ethyl ester, enamine_001390, ethyl 3-aminocoumarilate, ethyl3-aminobenzofuran-2-carboxylate | |
| ethyl 3-amino-1-benzofuran-2-carboxylate |
Specifications
| 39786-35-1 | |
| White-Yellow | |
| MFCD00641330 | |
| ethyl 3-aminobenzofuran-2-carboxylate, 3-aminocoumarilic acid ethyl ester, 2-benzofurancarboxylic acid, 3-amino-, ethyl ester, ethyl 3-aminobenzo b furan-2-carboxylate, ethyl 3-azanyl-1-benzofuran-2-carboxylate, 3-amino-2-benzofurancarboxylic acid ethyl ester, 3-aminobenzofuran-2-carboxylic acid ethyl ester, enamine_001390, ethyl 3-aminocoumarilate, ethyl3-aminobenzofuran-2-carboxylate | |
| CCOC(=O)C1=C(C2=CC=CC=C2O1)N | |
| 205.213 | |
| 205.21 | |
| Crystalline Powder |
| 80°C | |
| C11H11NO3 | |
| 1 g | |
| PWOARNMOPCOJEV-UHFFFAOYSA-N | |
| ethyl 3-amino-1-benzofuran-2-carboxylate | |
| 2063537 | |
| ≥98.0% (GC) | |
| Ethyl 3-Aminobenzofuran-2-carboxylate |
Safety and Handling
TSCA : No