missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminobenzofuran-2-carboxylate 98.0+%, TCI America™

Catalog No. E09101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09101G 1 g
E09105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09101G Supplier TCI America Supplier No. E09101G
Only null left

Chemical Identifiers

CAS 39786-35-1
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00641330
InChI Key PWOARNMOPCOJEV-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzofuran-2-carboxylate, 3-aminocoumarilic acid ethyl ester, 2-benzofurancarboxylic acid, 3-amino-, ethyl ester, ethyl 3-aminobenzo b furan-2-carboxylate, ethyl 3-azanyl-1-benzofuran-2-carboxylate, 3-amino-2-benzofurancarboxylic acid ethyl ester, 3-aminobenzofuran-2-carboxylic acid ethyl ester, enamine_001390, ethyl 3-aminocoumarilate, ethyl3-aminobenzofuran-2-carboxylate
PubChem CID 2063537
IUPAC Name ethyl 3-amino-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC=C2O1)N

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
Formula Weight 205.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Aminobenzofuran-2-carboxylate