missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-4,4,4-trifluorocrotonate, 97%

Catalog No. AAB2254506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2254506 5 g
AAB2254514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2254506 Supplier Thermo Scientific Chemicals Supplier No. B2254506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 372-29-2 | C6H8F3NO2 | 183.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-29-2
Molecular Formula C6H8F3NO2
Molecular Weight (g/mol) 183.13
MDL Number MFCD00068195
InChI Key NXVKRKUGIINGHD-ONEGZZNKSA-N
PubChem CID 10899311
IUPAC Name ethyl (E)-3-amino-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)C=C(C(F)(F)F)N

Specifications

Melting Point 24°C to 26°C
Density 1.245
Boiling Point 83°C to 85°C (15 mmHg)
Flash Point 65°C (149°F)
Odor Amine-like
Refractive Index 1.4229
Quantity 5 g
UN Number UN3259
Beilstein 4397839
Sensitivity Air Sensitive
Synonym ethyl 3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluorobut-2-enoate, ethyl 2z-3-amino-4,4,4-trifluorobut-2-enoate, 3-amino-4,4,4-trifluorocrotonic acid ethyl ester, ethyl-3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluoro-2-butenoate, ethyl z-3-amino-4,4,4-trifluorobut-2-enoate, ethyl z-3-amino-4,4,4-trifluoro-but-2-enoate, hcvdpjfpdddldlbdlrfuulu@bh, ethyl3-amino-4,4,4-trifluorocrotonate
Formula Weight 183.13
Percent Purity 97%
Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorocrotonate
Show More Show Less
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only