missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-4,4,4-trifluorobutyrate, 95%, Thermo Scientific Chemicals

Catalog No. AAL1702503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1702503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1702503 Supplier Thermo Scientific Chemicals Supplier No. L1702503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 170804-18-9 | Molecular Formula: C6H10F3NO2 | Molecular Weight: 185.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170804-18-9
Molecular Formula C6H10F3NO2
Molecular Weight (g/mol) 185.146
MDL Number MFCD01075754
InChI Key KGUOZZWGWBBRRF-UHFFFAOYSA-N
Synonym ethyl 3-amino-4,4,4-trifluorobutyrate, ethyl-3-amino-4,4,4-trifluorobutyrate, acmc-20akzx, ethyl3-amino-4,4,4-trifluorobutyrate, 3-amino-4,4,4-trifluorobutyric acid ethyl ester
PubChem CID 2774311
IUPAC Name ethyl 3-amino-4,4,4-trifluorobutanoate
SMILES CCOC(=O)CC(C(F)(F)F)N

Specifications

Boiling Point 70°C to 71°C (4mmHg)
Refractive Index 1.3831
UN Number UN2735
Beilstein 6136102
Quantity 1 g
Formula Weight 185.15
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorobutyrate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only