missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-1H-pyrazole-4-carboxylate, 99%

Catalog No. p-7081066
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1088206 5 g
AAA1088214 25 g
AAA1088222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1088206 Supplier Thermo Scientific Chemicals Supplier No. A1088206
Only null left

CAS: 6994-25-8 | C6H9N3O2 | 155.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6994-25-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00005238
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
PubChem CID 81472
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)N

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 4964
Formula Weight 155.16
Percent Purity 99%
Chemical Name or Material Ethyl 3-amino-1H-pyrazole-4-carboxylate
EINECSNumber 230-262-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only