missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-Dimethylisoxazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E12121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12121G 1 g
E12125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12121G Supplier TCI America Supplier No. E12121G

Chemical Identifiers

CAS 17147-42-1
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00052557
InChI Key OQRHVDXUJAQVNT-UHFFFAOYSA-N
Synonym ethyl 3,5-dimethylisoxazole-4-carboxylate, ethyl 3,5-dimethyl-4-isoxazolecarboxylate, 3,5-dimethylisoxazole-4-carboxylic acid ethyl ester, 4-isoxazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3,5-dimethyl-4-ethoxycarbonyl isoxazole, ethyl 3,5-dimethyl-isoxazole-4-carboxylate, acmc-1c48c, ethyldimethylisoxazolecarboxylate, 4-carbethoxy-3,5-dimethylisoxazole, 4-carboethoxy-3,5-dimethylisoxazole
PubChem CID 736348
IUPAC Name ethyl 3,5-dimethyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=C(ON=C1C)C

Specifications

Color Yellow
Boiling Point 220°C
Quantity 1 g
Formula Weight 169.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3,5-Dimethylisoxazole-4-carboxylate