missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dimethylisoxazole-4-carboxylate, 97%

Catalog No. p-7025252
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2064288 2.5 g
AAB2064209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2064288 Supplier Thermo Scientific Chemicals Supplier No. B2064288
Only null left

CAS: 17147-42-1 | C8H11NO3 | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17147-42-1
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00052557
InChI Key OQRHVDXUJAQVNT-UHFFFAOYSA-N
Synonym ethyl 3,5-dimethylisoxazole-4-carboxylate, ethyl 3,5-dimethyl-4-isoxazolecarboxylate, 3,5-dimethylisoxazole-4-carboxylic acid ethyl ester, 4-isoxazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3,5-dimethyl-4-ethoxycarbonyl isoxazole, ethyl 3,5-dimethyl-isoxazole-4-carboxylate, acmc-1c48c, ethyldimethylisoxazolecarboxylate, 4-carbethoxy-3,5-dimethylisoxazole, 4-carboethoxy-3,5-dimethylisoxazole
PubChem CID 736348
IUPAC Name ethyl 3,5-dimethyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=C(ON=C1C)C

Specifications

Boiling Point 87°C to 89°C (2 mmHg)
Quantity 2.5 g
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material Ethyl 3,5-dimethylisoxazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only