missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate 98.0+%, TCI America™

Catalog No. E08435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08435G 5 g
E084325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08435G Supplier TCI America Supplier No. E08435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2199-44-2
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00010645
InChI Key IZSBSZYFPYIJDI-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 3,5-dimethylpyrrole-2-carboxylate, ethyl 3,5-dimethyl-2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester, 3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester, ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate, pubchem23984, acmc-1cdc7, 3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester, ksc496q7t
PubChem CID 137477
IUPAC Name ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(N1)C)C

Specifications

Melting Point 124°C
Boiling Point 272°C
Quantity 5 g
Formula Weight 167.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate