missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-Bis(2-methoxyethoxy)benzoate 98.0+%, TCI America™

Catalog No. E11865G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11865G 5 g
E11861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11865G Supplier TCI America Supplier No. E11865G
Only null left

Chemical Identifiers

CAS 183322-16-9
Molecular Formula C15H22O6
Molecular Weight (g/mol) 298.335
MDL Number MFCD01935284
InChI Key VGFZRAVMWXHEJB-UHFFFAOYSA-N
Synonym 3,4-Bis(2-methoxyethoxy)benzoic Acid Ethyl Ester
PubChem CID 2808992
IUPAC Name ethyl 3,4-bis(2-methoxyethoxy)benzoate
SMILES CCOC(=O)C1=CC(=C(C=C1)OCCOC)OCCOC

Specifications

Melting Point 56°C
Color White
Quantity 5 g
Formula Weight 298.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,4-Bis(2-methoxyethoxy)benzoate