missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trimethoxybenzoate, 98%

Catalog No. AAA1319906
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319906 5 g
AAA1319922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1319906 Supplier Thermo Scientific Chemicals Supplier No. A1319906
Only null left

CAS: 6178-44-5 | C12H16O5 | 240.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6178-44-5
Molecular Formula C12H16O5
Molecular Weight (g/mol) 240.255
MDL Number MFCD00017525
InChI Key UEOFNBCUGJADBM-UHFFFAOYSA-N
PubChem CID 231162
IUPAC Name ethyl 3,4,5-trimethoxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC

Specifications

Quantity 5 g
Synonym 3,4,5-trimethoxybenzoic acid ethyl ester, ethyl 3,4,5-trimethoxy benzoate, ethyltrimethoxy benzoate, acmc-1b7mp, ethyl 3,4,5-trimethoxybenzoate;, benzoic acid,3,4,5-trimethoxy-, ethyl ester, benzoic acid, 3,4,5-trimethoxy-, ethyl ester
Formula Weight 240.26
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trimethoxybenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only