missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trihydroxybenzoate, 98%

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol

$94.08 - $278.92

Chemical Identifiers

Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1373309
View Documents
Thermo Scientific Chemicals
A1373309
10 g
Each for $94.08
Only null left
 
AAA1373318
View Documents
Thermo Scientific Chemicals
A1373318
50 g
Each for $278.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

C9H10O5
MFCD00016430
ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
CHEBI:87247
CCOC(=O)C1=CC(=C(C(=C1)O)O)O
198.174
VFPFQHQNJCMNBZ-UHFFFAOYSA-N
13250
ethyl 3,4,5-trihydroxybenzoate

Specifications

148°C to 152°C
MFCD00016430
ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
CCOC(=O)C1=CC(=C(C(=C1)O)O)O
198.174
CHEBI:87247
0.98
C9H10O5
10 g
VFPFQHQNJCMNBZ-UHFFFAOYSA-N
ethyl 3,4,5-trihydroxybenzoate
13250
198.18
Ethyl 3,4,5-trihydroxybenzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-608-5

RTECSNumber : LW7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only