missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trihydroxybenzoate, 98%

Catalog No. AAA1373309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1373309 10 g
AAA1373318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1373309 Supplier Thermo Scientific Chemicals Supplier No. A1373309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 148°C to 152°C
Quantity 10 g
Formula Weight 198.18
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trihydroxybenzoate
EINECSNumber 212-608-5
RTECSNumber LW7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only