missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-Diethoxypropionate 95.0+%, TCI America™

Catalog No. D196225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D196225ML 25 mL
D1962100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D196225ML Supplier TCI America Supplier No. D196225ML

Chemical Identifiers

CAS 10601-80-6
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.239
MDL Number MFCD00009865
InChI Key SIALOQYKFQEKOG-UHFFFAOYSA-N
Synonym ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
PubChem CID 66389
IUPAC Name ethyl 3,3-diethoxypropanoate
SMILES CCOC(CC(=O)OCC)OCC

Specifications

Color Colorless
Boiling Point 86°C
Quantity 25 mL
Formula Weight 190.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3,3-Diethoxypropionate