missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-diethoxypropionate, 96%

Catalog No. p-7045140
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0956209 10 g
AAL0956218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0956209 Supplier Thermo Scientific Chemicals Supplier No. L0956209
Only null left

CAS: 10601-80-6 | C9H18O4 | 190.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10601-80-6
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.239
MDL Number MFCD00009865
InChI Key SIALOQYKFQEKOG-UHFFFAOYSA-N
PubChem CID 66389
IUPAC Name ethyl 3,3-diethoxypropanoate
SMILES CCOC(CC(=O)OCC)OCC

Specifications

Density 0.978
Boiling Point 92°C to 93°C (13 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.412
Quantity 10 g
Beilstein 1772110
Synonym ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
Formula Weight 190.24
Percent Purity 96%
Chemical Name or Material Ethyl 3,3-diethoxypropionate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 234-223-1
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only