missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 269641000

Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Catalog No. AC269641000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10601-80-6
190.24
SIALOQYKFQEKOG-UHFFFAOYSA-N
66389
CCOC(CC(=O)OCC)OCC
C9H18O4
MFCD00009865
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
ethyl 3,3-diethoxypropanoate

Specifications

Ethyl 3, 3-diethoxypropanoate
0.9700g/mL
93°C
98%
C9H18O4
(C2H5O)2CHCH2COOC2H5
MFCD00009865
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
SIALOQYKFQEKOG-UHFFFAOYSA-N
ethyl 3,3-diethoxypropanoate
66389
98%
10601-80-6
95°C (17.0mmHg)
Authentic
Glass bottle
1.4100 to 1.412
100mL
0.97
Solubility in water: 23g/L (20 c)
CCOC(CC(=O)OCC)OCC
190.24
190.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-223-1

SDS
Documents

Documents

Promotions

Promotions