missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 269640050
| Quantity | 5mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10601-80-6 | |
| 190.24 | |
| SIALOQYKFQEKOG-UHFFFAOYSA-N | |
| 66389 | |
| CCOC(CC(=O)OCC)OCC |
| C9H18O4 | |
| MFCD00009865 | |
| ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate | |
| ethyl 3,3-diethoxypropanoate |
Specifications
| Ethyl 3, 3-diethoxypropanoate | |
| 0.9700g/mL | |
| 93°C | |
| 98% | |
| C9H18O4 | |
| (C2H5O)2CHCH2COOC2H5 | |
| MFCD00009865 | |
| ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate | |
| SIALOQYKFQEKOG-UHFFFAOYSA-N | |
| ethyl 3,3-diethoxypropanoate | |
| 66389 | |
| 98% |
| 10601-80-6 | |
| 95°C (17.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4100 to 1.412 | |
| 5mL | |
| 0.97 | |
| Solubility in water: 23g/L (20 c) | |
| CCOC(CC(=O)OCC)OCC | |
| 190.24 | |
| 190.24 |
Safety and Handling
EINECSNumber : 234-223-1