missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-Bis(methylthio)-2-cyanoacrylate 98.0+%, TCI America™

Catalog No. B141125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B141125G 25 g
B1411500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B141125G Supplier TCI America Supplier No. B141125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17823-58-4
Molecular Formula C8H11NO2S2
Molecular Weight (g/mol) 217.30
MDL Number MFCD00143027
InChI Key POABRARINOCORV-UHFFFAOYSA-N
Synonym 3,3-Bis(methylthio)-2-cyanoacrylic Acid Ethyl Ester
PubChem CID 2747745
IUPAC Name ethyl 2-cyano-3,3-bis(methylsulfanyl)prop-2-enoate
SMILES CCOC(=O)C(C#N)=C(SC)SC

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 25 g
Formula Weight 217.30
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,3-Bis(methylthio)-2-cyanoacrylate