missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate 98.0+%, TCI America™
$48.46 - $160.30
Chemical Identifiers
| CAS | 212322-56-0 |
|---|---|
| Molecular Formula | C18H22N4O3 |
| Molecular Weight (g/mol) | 342.399 |
| MDL Number | MFCD09833624 |
| InChI Key | PCPATNZTKBOKOY-UHFFFAOYSA-N |
| Synonym | 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester |
| PubChem CID | 11982993 |
| IUPAC Name | ethyl 3-[[3-amino-4-(methylamino)benzoyl]-pyridin-2-ylamino]propanoate |
| SMILES | CCOC(=O)CCN(C1=CC=CC=N1)C(=O)C2=CC(=C(C=C2)NC)N |
Chemical Identifiers
| 212322-56-0 | |
| 342.399 | |
| PCPATNZTKBOKOY-UHFFFAOYSA-N | |
| 11982993 | |
| CCOC(=O)CCN(C1=CC=CC=N1)C(=O)C2=CC(=C(C=C2)NC)N |
| C18H22N4O3 | |
| MFCD09833624 | |
| 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester | |
| ethyl 3-[[3-amino-4-(methylamino)benzoyl]-pyridin-2-ylamino]propanoate |
Specifications
| 212322-56-0 | |
| White-Yellow | |
| MFCD09833624 | |
| 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester | |
| CCOC(=O)CCN(C1=CC=CC=N1)C(=O)C2=CC(=C(C=C2)NC)N | |
| 342.399 | |
| 342.40 | |
| Crystalline Powder |
| 106°C | |
| C18H22N4O3 | |
| 1 g | |
| PCPATNZTKBOKOY-UHFFFAOYSA-N | |
| ethyl 3-[[3-amino-4-(methylamino)benzoyl]-pyridin-2-ylamino]propanoate | |
| 11982993 | |
| ≥98.0% (HPLC,T) | |
| Ethyl 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator