missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate 98.0+%, TCI America™

Catalog No. E10331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10331G 1 g
E10335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10331G Supplier TCI America Supplier No. E10331G

Chemical Identifiers

CAS 212322-56-0
Molecular Formula C18H22N4O3
Molecular Weight (g/mol) 342.399
MDL Number MFCD09833624
InChI Key PCPATNZTKBOKOY-UHFFFAOYSA-N
Synonym 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester
PubChem CID 11982993
IUPAC Name ethyl 3-[[3-amino-4-(methylamino)benzoyl]-pyridin-2-ylamino]propanoate
SMILES CCOC(=O)CCN(C1=CC=CC=N1)C(=O)C2=CC(=C(C=C2)NC)N

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 1 g
Formula Weight 342.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate
TSCA No
Recommended Storage Refrigerator