missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-furyl)propionate, 98%

Catalog No. p-7024310
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1873206 5 g
AAA1873214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1873206 Supplier Thermo Scientific Chemicals Supplier No. A1873206

CAS: 10031-90-0 | C9H12O3 | 168.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10031-90-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00036496
InChI Key OWIWZQQFSTZZIG-UHFFFAOYSA-N
Synonym ethyl 3-2-furyl propionate, ethyl 2-furanpropionate, 2-furanpropanoic acid, ethyl ester, ethyl 3-2-furyl propanoate, ethyl furfhydracrylate, ethyl furfurylacetate, ethyl 3-furan-2-yl propanoate, 2-furanpropionic acid, ethyl ester, ethyl 2-furanpropanoate, ethyl furan-2-propionate
PubChem CID 61450
IUPAC Name ethyl 3-(furan-2-yl)propanoate
SMILES CCOC(=O)CCC1=CC=CO1

Specifications

Quantity 5 g
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material Ethyl 3-(2-furyl)propionate
EINECSNumber 233-097-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only