missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog No. p-7028462
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2579106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2579106 Supplier Thermo Scientific Chemicals Supplier No. H2579106
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33545-44-7
Molecular Formula C10H10N2O3
Molecular Weight (g/mol) 206.20
MDL Number MFCD09953454,MFCD06797478
InChI Key GSVBXAZEQDJUOY-UHFFFAOYSA-N
Synonym ethyl 5-2-furyl-1h-pyrazole-3-carboxylate, ethyl 5-furan-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-furan-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-fur-2-yl-1h-pyrazole-3-carboxylate, ethyl 5-2-furyl pyrazole-3-carboxylate, ethyl 5-furan-2-yl-2h-pyrazole-3-carboxylate, aronis23991, ethyl 3-2-furyl pyrazole-5-carboxylate, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-, ethyl ester
PubChem CID 7127811
IUPAC Name ethyl 3-(furan-2-yl)-1H-pyrazole-5-carboxylate
SMILES CCOC(=O)C1=CC(=NN1)C1=CC=CO1

Specifications

Melting Point 135°C to 137°C
Quantity 5 g
Formula Weight 206.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only