missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-thiouracil-5-carboxylate, 99%

Catalog No. AAA1704606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1704606 5 g
AAA1704614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1704606 Supplier Thermo Scientific Chemicals Supplier No. A1704606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38026-46-9 | C7H8N2O3S | 200.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38026-46-9
Molecular Formula C7H8N2O3S
Molecular Weight (g/mol) 200.21
MDL Number MFCD00006074
InChI Key FQFSHLBWRUOCPX-UHFFFAOYSA-N
Synonym 5-carbethoxy-2-thiouracil, ethyl 2-thiouracil-5-carboxylate, 2-mercapto-5-carbethoxyuracil, 5-ethoxycarbonyl-2-thiouracil, uracil, 5-carbethoxy-2-thio, ethyl 4-hydroxy-2-mercaptopyrimidine-5-carboxylate, 5-carboethoxy-2-thiouracil, ethyl 4-oxo-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-4-oxo-2-thio-, ethyl ester, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-4-oxo-2-thioxo-, ethyl ester
PubChem CID 735952
IUPAC Name ethyl 4-oxo-2-sulfanylidene-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CNC(=S)NC1=O

Specifications

Melting Point ∼250°C (decomposition)
Quantity 5 g
Beilstein 190867
Formula Weight 200.22
Percent Purity 99%
Chemical Name or Material Ethyl 2-thiouracil-5-carboxylate
EINECSNumber 253-749-2
RTECSNumber UV7775000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only