missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(p-Toluenesulfonyloxy)acetate 98.0+%, TCI America™

Catalog No. p-7129183
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08511G 1 g
E08515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08511G Supplier TCI America Supplier No. E08511G

Chemical Identifiers

CAS 39794-75-7
Molecular Formula C11H14O5S
Molecular Weight (g/mol) 258.29
MDL Number MFCD18252874
InChI Key CVNRARCXNKDWKI-UHFFFAOYSA-N
Synonym 2-(p-Toluenesulfonyloxy)acetic Acid Ethyl Ester
PubChem CID 11821264
IUPAC Name ethyl 2-[(4-methylbenzenesulfonyl)oxy]acetate
SMILES CCOC(=O)COS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 47°C
Color White
Quantity 1 g
Formula Weight 258.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(p-Toluenesulfonyloxy)acetate