missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH5790603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5790603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5790603 Supplier Thermo Scientific Chemicals Supplier No. H5790603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Recommended storage temperature: 2 - 8°C Handle and store under inert gas.

Chemical Identifiers

CAS 36821-26-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD06796534
InChI Key FHHMWJMUMOYGQW-UHFFFAOYSA-N
PubChem CID 12699000
IUPAC Name ethyl 2-oxopyrrolidine-3-carboxylate
SMILES CCOC(=O)C1CCNC1=O

Specifications

Melting Point 71°C to 75°C
Quantity 1 g
Sensitivity Light sensitive
Synonym 2-oxo-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 2-oxo-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 2-oxo-, ethyl ester, 2-oxopyrrolidine-3-carboxylic acid ethyl ester, 3-carboethoxy-pyrrolidinone, ethyl2-oxo-pyrrolidine-3-carboxylate, ethyl 2-oxopyrrolidine-3-carboxylate, n-boc-2-oxo-d-beta-proline ethyl ester, 3-pyrrolidinecarboxylicacid, 2-oxo-, ethyl ester
Solubility Information Slightly soluble in water.
Formula Weight 157.17
Percent Purity 95%
Chemical Name or Material Ethyl 2-oxopyrrolidine-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only