missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Oxocyclopentanecarboxylate 97.0+%, TCI America™

Catalog No. B026825ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B026825ML 25 mL
B0268250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B026825ML Supplier TCI America Supplier No. B026825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-10-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00001412
InChI Key JHZPNBKZPAWCJD-UHFFFAOYSA-N
Synonym ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
PubChem CID 69136
IUPAC Name ethyl 2-oxocyclopentane-1-carboxylate
SMILES CCOC(=O)C1CCCC1=O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
Formula Weight 156.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Oxocyclopentanecarboxylate