missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclopentanecarboxylate, 97+%

CAS: 611-10-9 | C8H12O3 | 156.181 g/mol

Supplier:  Thermo Scientific Chemicals L0452822

 View more versions of this product

Catalog No. AAL0452822


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 2-oxocyclopentanecarboxylate
1.079
77°C (170°F)
1.452
100 g
ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
CCOC(=O)C1CCCC1=O
156.181
156.18
611-10-9
225°C to 228°C
C8H12O3
MFCD00001412
387787
JHZPNBKZPAWCJD-UHFFFAOYSA-N
ethyl 2-oxocyclopentane-1-carboxylate
69136
≥97%

Chemical Identifiers

611-10-9
156.181
JHZPNBKZPAWCJD-UHFFFAOYSA-N
69136
CCOC(=O)C1CCCC1=O
C8H12O3
MFCD00001412
ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
ethyl 2-oxocyclopentane-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-253-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only